Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonaqueous Buffer Stock Solution B, Ricca Chemical
SDP

Catalog No. 546316 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5463-16 500 mL
5463-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5463-16 Supplier Ricca Chemical Company Supplier No. 546316
Add to Cart
Add to Cart

For Acid Number and Neutralization Number of Petroleum Products

Specifications

Chemical Name or Material Nonaqueous Buffer Stock Solution B
Name Note For Acid Number and Neutralization Number of Petroleum Products
CAS 67-63-0, 554-84-7, 1310-58-3, 107-15-3
CAS Min % 94.0, 3.33, 0.4, 0.02
Color Amber
CAS Max % 98.0, 3.47, 0.41, 0.02
Concentration 1X
For Use With (Application) Water Analysis
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Packaging Amber Glass Bottle
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.