Learn More
Enzo Life Sciences Nonivamide (100mg). CAS: 2444-46-4
SDP
Supplier:  Enzo Life Sciences ALX550239M100
Encompass_Preferred
Minor constituent of hot pepper (Capsicum oleoresin). Alternative name: Nonylic vanillylamide, N-[4-Hydroxy-3-methoxy-benzyl]nonanamide, N-Vanillylnonanamide, Pelargonic acid vanillylamide, Vanillyl pelargonic amide, Pseudocapsaicin. Purity: ≥96% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO or 100% ethanol; slightly soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9506
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.