Learn More
Covidien Nonsterile Monoject™ Syringes without Needles
Catalog No. 22257143
Change view
Click to view available options
Volume (Metric):
3 mL
6 mL
12 mL
Connection Type:
Regular
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Connection Type
22-257-143 3 mL Regular
22-257-144 6 mL Regular
22-257-145 12 mL Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-257-143 Supplier Covidien Supplier No. 8881103025
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Bulk packaged without needles, available in several sizes
  • Permanent heat-etched graduations allow precise fluid measurements
  • Self-sealing, latex-free plunger tip
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Used as general purpose syringe
Latex Free Yes
Product Type Monoject™ Syringe
Needle Point Number No Needle Included
Volume (Metric) 3 mL
Connection Type Regular
Graduated Yes
Sterility Non-sterile