Learn More
Nonylphenol monoethoxylate Solution, 5 μg/mL in Acetone, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 67-64-1 |
| Flash Point | -17°C - Closed Cup |
| Synonym | NP-1 EO technical |
| Type | Analytical Standard |
| Quantity | 10 mL |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 5 μg/mL in acetone |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.