Learn More
Apexbio Technology LLC ML-354 89159-60-4 25mg
Supplier: Apexbio Technology LLC C305825
ML-354 (CAS 89159-60-4) is an indole-derived antagonist of protease-activated receptor 4 (PAR4) exhibiting an IC50 of 140 nM PAR4 is a G-protein-coupled receptor activated via proteolytic cleavage by serine proteases and serves as a thrombin receptor on human platelets By inhibiting PAR4 ML-354 provides a tool to investigate PAR4-mediated platelet activation pathways Its selectivity over PAR1 supports its use in the study of thrombotic mechanisms and the evaluation of PAR4 as a therapeutic target in platelet-related disorders Current research is focused on the compound s in vitro and in vivo pharmacological characterization
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.