Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Norcross Safety PVC Pull-On Boots

Specifications

Certifications/Compliance ASTM F2413-11 M I/75 C/75
Color Black
For Use With (Application) General industrial, farming, general occupation
Height (English) 15 in.
Material PVC
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-051813
View Documents
Honeywell™
18801BLM080
8 N/A N/A N/A
Description

Description

Specifications

Specifications

ASTM F2413-11 M I/75 C/75
General industrial, farming, general occupation
PVC
Steel
Steel
Yes
Black
15 in.
Seamless
Deep-angle cleated sole
Degradation from agricultural chemicals, fertilizers and animal waste
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.