Learn More
U.S. Pharmacopeia Norepinephrine Bitartrate, 108341-18-0, MFCD00036384, 125mg
Supplier:  U.S. Pharmacopeia 1468501
Encompass
Molecular formula C8H11NO3.C4H6O6.H2O, Molecular weight 337.28, Melting point/freezing point 208.4 - 219.2 °F (98 - 104 °C), pH in aqueous solution 3.5, Synonyms: Noradrenaline bitartrate monohydrate, Levarterenol bitartrate monohydrate, 1,2-Benzenediol, 4-(2-amino-1-hydroxyethyl)-, (R)-[R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), monohydrate
Catalog No. NC1354172
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.