Learn More
Alpco Ltd Norepinephrine ELISA
SDP
Supplier:  Alpco Ltd 17NORHUE01.1
Encompass_Preferred
The Norepinephrine ELISA is an enzyme immunoassay for the quantitative determination of Noradrenaline (Norepinephrine) in plasma and urine. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3496
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.