Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Norepinephrine Transporter Monoclonal Antibody (CL3063)
GREENER_CHOICE

Catalog No. PIMA524647 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524647 Supplier Invitrogen™ Supplier No. MA524647
  • $689.00
    $506.65 / Each of 1

    Save $182.35

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Immunogen sequence: SLYYLFSSFT LNLPWTDCGH TWNSPNCTDP KLLNGSVLGN HTKYSKYKFT PAAEFYERGV LHLHESSGIH DI Binds to an epitope located within the peptide sequence HTKYSKYKFTPAAEF as determined by overlapping synthetic peptides. Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 93%.

Norepinephrine Transporter [NET, or noradrenaline transporter (NAT)] is a monoamine transporter that transports the neurotransmitter noradrenaline from the synapse back to its vesicles for storage until later use. It also appears to transport the neurotransmitter dopamine in the same way, but to a lesser degree. Studies have shown a decrease in NET levels in the locus coeruleus in patients diagnosed with major depression. Cocaine, amphetamines and many therapeutic antidepressants, such as the SNRIs (serotonin-norepinephrine reuptake inhibitors) and the tricyclic antidepressants (TCAs) act to raise noradrenaline. Furthermore, deficits in the NET gene have been associated with ADHD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Norepinephrine Transporter
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL3063
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC6A2
Gene Accession No. O55192, P23975
Gene Alias NaCl-dependent norepinephrine transporter; NAT1; NE transporter; NET; NE-T; NET1; neurotransmitter transporter; neurotransmitter transporter, noradrenal; NORADR; noradrenaline; noradrenaline transporter; norepinephrine transporter; Slc6a2; SLC6A5; sodium-dependent noradrenaline transporter; solute carrier family 6 (neurotransmitter transporter), member 2; solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2; solute carrier family 6 member 2; solute carrier family 6 member 5; transmembrane region 4..26 transmembrane region 55..77 transmembrane region 105..123 transmembrane region 136..157
Gene Symbols SLC6A2
Host Species Mouse
Immunogen Recombinant protein fragment of Human SLC6A2 (Product #RP-102393)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20538, 6530, 83511
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.