Learn More
MyBioSource Polyclonal Norfentanyl Antibody
SDP
Supplier:  MyBioSource MBS3408551MG
Encompass_Preferred
The MyBioSource Polyclonal Norfentanyl Antibody is a rabbit polyclonal antibody raised against norfentanyl conjugated to KLH. It is recommended for detection of norfentanyl and its metabolites in human serum, plasma, tissue homogenates or other biological fluids. The antibody has high affinity and can detect levels as low as 0.1 ng/ml in ELISA. This antibody can be used in ELISA, WB and IHC. It is provided at 100μg in 1mg/ml of PBS with 0.02% sodium azide and 0.5% BSA.
Catalog No. NC2880671
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.