Learn More
Abcam Norflo x acin, Antibacterial agent, 1G
Supplier: Abcam AB1412191G
MW 319.33 Da, Purity >99%. Broad spectrum antibacterial agent active against Gram-positive and Gram-negative bacteria. Inhibits cell division by inhibiting DNA gyrase. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.