Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Norfloxacin, G-Biosciences
SCI_ED

Catalog No. S139822
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139822 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139822 Supplier G-Biosciences Supplier No. RC1071
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Norfloxacin is a derivative of pefloxacin.

Norfloxacin is a first generation fluoroquinolone antibiotic targeting primarily gram-negative bacteria responsible for causing urinary tract infections

Chemical Identifiers

CAS 70458-96-7
Molecular Weight (g/mol) 319.33 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 10 g
Percent Purity >98%
Grade Molecular Biology
Chemical Name or Material Norfloxacin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.