accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHOOFNNH

CAS RN 70458-96-7

IUPAC Name: 1-ethyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid
Synonyms: norfloxacin sebercim norfloxacin (substance) NFLX norfloxacine norfloxacino norfloxacinum norfloxacin (product) barazan fulgram
Molecular Weight (g/mol): 319.34
Molecular Formula: C16H18FN3O3
InChi Key: OGJPXUAPXNRGGI-UHFFFAOYSA-N
SMILES: CCN1C=C(C(O)=O)C(=O)C2=CC(F)=C(C=C12)N1CCNCC1
Molecular Weight (g/mol) 
  • (1)
  • (7)
Physical Form 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (1)
Grade 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (7)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)