Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Norfloxacin, MP Biomedicals

Catalog No. ICN15594925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15594925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15594925 Supplier MP Biomedicals, Inc Supplier No. 0215594925
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 70458-96-7
Molecular Formula C16H18FN3O3
Molecular Weight (g/mol) 319.336
InChI Key OGJPXUAPXNRGGI-UHFFFAOYSA-N
PubChem CID 4539
ChEBI CHEBI:100246
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O

Specifications

Physical Form Powder
Quantity 25 g
Grade Not Available
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.