Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Goat IgG, Negative Control, R&D Systems™
GSA_VA

Catalog No. AB108C
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB108C 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB108C Supplier R&D Systems Supplier No. AB108C
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 282 publications

IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Control.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen IgG
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Control
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Goat
Immunogen NA
Purification Method Protein A or G purified
Quantity 1 mg
Gene ID (Entrez) 3500
Test Specificity Serum was obtained from naive (non-immunized) goats and purified for use as normal goat IgG.
Reconstitution Dissolve the lyophilized goat IgG in sterile PBS, pH 7.4. If 1 mL of PBS is used, the antibody concentration will be 1 mg/mL.
Target Species Goat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.