Learn More
Normal Human IgG, Isotype Control, R&D Systems™
Description
Specifications
Specifications
| Antigen | IgG |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Control |
| Formulation | 10 mM Sodium Phosphate, 0.85% (w/v) Sodium Chloride, pH 7.2 (PBS) with 5% trehalose and 0.05% (w/v) Sodium Azide with No Preservative |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Gene Symbols | IGHG1 |
| Host Species | Human |
| Immunogen | Normal human plasma (Donor plasma tested and found negative for HBsAg, HIV-1, HIV-2 and HCV antibodies by FDA approved tests) |
| Purification Method | Protein A or G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.