Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Mouse IgM, Alexa Fluor 488, Negative Control, R&D Systems™

Catalog No. IC015G
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC015G 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC015G Supplier R&D Systems Supplier No. IC015G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Polyclonal Antibody

IgM Polyclonal specifically detects IgM in Mouse samples. It is validated for Control.

Specifications

Antigen IgM
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Control 5 uL/10^6 cells
Gene Alias AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Gene Symbols IGHM
Host Species Mouse
Immunogen NA
Purification Method Protein A or G purified
Quantity 200 Tests
Gene ID (Entrez) 3507
Test Specificity Serum was obtained from naive (non-immunized) mice and purified for use as a mouse IgM control for flow cytometry applications.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.