Learn More
Normal Rabbit IgG, Alexa Fluor™ 488 conjugate Rabbit, AlexaFluor™488, Polyclonal, Isotype Control, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | IgG |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Host Species | Rabbit |
| Purification Method | Fractionation and ion exchange chromatography |
| Quantity | 50 μg |
| Primary or Secondary | Primary |
| Form | Purified |
| Isotype | IgG |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.