Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Rabbit IgG, Negative Control, R&D Systems™
SDP

Catalog No. AB105C
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB105C 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB105C Supplier R&D Systems Supplier No. AB105C

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 64 publications

IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Control.

Specifications

Antigen IgG
Applications Control
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Control
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Rabbit
Immunogen NA
Purification Method Protein A or G purified
Quantity 1 mg
Regulatory Status RUO
Gene ID (Entrez) 3500
Test Specificity Serum was obtained from naive (non-immunized) rabbits and purified for use as normal rabbit IgG.
Reconstitution Dissolve the lyophilized rabbit IgG in sterile PBS, pH 7.4. If 1 mL of PBS is used, the antibody concentration will be 1 mg/mL.
Target Species Rabbit
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.