Learn More
Santa Cruz Biotechnology NORMAL RAT IGG
Supplier:  Santa Cruz Biotechnology SC2026
Encompass
Normal rat IgG; Use as negative control for Western blotting, immunoprecipitation and immunohistochemistry applications; Storage: 4 deg.C; 200ug/0.5mL
Catalog No. NC9874065
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.