Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Rat IgG, Isotype Control (Azide Free), R&D Systems™

Catalog No. 6001F
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001F 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 6001F Supplier R&D Systems Supplier No. 6001F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Polyclonal Antibody has been used in 3 publications

IgG Polyclonal specifically detects IgG in Rat samples. It is validated for Control.

Specifications

Antigen IgG
Classification Polyclonal
Conjugate Unconjugated
Dilution Control
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Rat
Quantity 1 mg
Gene ID (Entrez) 3500
Target Species Rat
Content And Storage Store the unopened product at 2 - 8degrees C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.