Learn More
Normal Sheep IgG, Alexa Fluor 488, Negative Control, R&D Systems™
Description
Specifications
Specifications
| Antigen | IgG |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Dilution | Control 5 uL/10^6 cells |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Gene Symbols | IGHG1 |
| Host Species | Sheep |
| Immunogen | NA |
| Purification Method | Protein A or G purified |
| Quantity | 200 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.