Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Normal Sheep IgG, Alexa Fluor 700, Negative Control, R&D Systems™
SDP

Catalog No. IC016N Shop All Novus Products
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC016N 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC016N Supplier R&D Systems Supplier No. IC016N

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Control.

Specifications

Antigen IgG
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Control 5 uL/10^6 cells
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Sheep
Immunogen NA
Purification Method Protein A or G purified
Quantity 200 Tests
Regulatory Status RUO
Gene ID (Entrez) 3500
Test Specificity Serum was obtained from naive (non-immunized) sheep.
Target Species Sheep
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.