Learn More
Invivogen Normocin™ - Antimicrobial Reagent, 1 g (1 x 20 ml)
Supplier: Invivogen ANTNR2
1 x 20mL. Product concentration: 50 mg/ml. Sterile water soluble solution. Active concentration not toxic for treated cell lines: 100 ug/ml. 1 ml Normocin is enough to treat 500 ml medium. Shipped at room temperature.