Learn More
Bioss KD-Validated NEK2 Rabbit Recom
Supplier: Bioss BSM90016R
NIMA Related Kinase 2 Primary Antibody Unconjugated Public Immunogen Range 21-47/445 Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.