Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ 4000 Replacement Tyvek Hood

Catalog No. 17640
Change view
Click to view available options
Hood; Clear Acetate Lens
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-640 Hood; Clear Acetate Lens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-640 Supplier Honeywell™ Supplier No. 14900409
This item is not returnable. View return policy

Specifications

Certifications/Compliance NIOSH
Color Clear
Material HDPE
Product Type Respirator Hood
For Use With (Equipment) Continuous flow supplied air hood
Includes Single bib, acetate lens
Model 14900409
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.