Learn More
Honeywell™ North™ A400 Series Safety Glasses
Sporty, lightweight safety glasses with one-piece wraparound lens
Specifications
Certifications/Compliance Meet ANSI Z87.1-2003 and CSA Z94.3 standards.
View More Specs
Products 4
Catalog Number Mfr. No. Lens Tint Frame Color Lens Coating Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Frame Color Lens Coating Price Quantity & Availability  
19-053-701B
View Documents
Honeywell™
A401
Gray Gray N/A N/A N/A
19-053-701A
View Documents
Honeywell™
A400
Clear Clear Scratch-resistant N/A N/A N/A
19-053-701D
View Documents
Honeywell™
A400AF
Clear Clear N/A N/A N/A
19-053-701E
View Documents
Honeywell™
A404
Silver, Indoor/Outdoor Gray Scratch-resistant N/A N/A N/A
Description
Description
  • Single lens and frame made of impact-resistant polycarbonate
  • Comfortable, wraparound design provides excellent peripheral vision protection without the need for sideshields
  • Feature antifog coating
Compliance
Meet ANSI™ Z87.1-2003 and CSA Z94.3 standards.
Specifications
Specifications
Meet ANSI Z87.1-2003 and CSA Z94.3 standards.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.