Learn More
Honeywell™ The Peak A79 Hard Hats
Catalog No. 19042024
Change view
Click to view available options
Color:
Orange
Red
Yellow
Suspension Type:
Pin-Lock
Ratchet
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-042-024 Orange Pin-Lock
19-042-025 Red Pin-Lock
19-042-027 Yellow Pin-Lock
19-042-056 Yellow Ratchet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-042-024 Supplier Honeywell™ Supplier No. A79030000
This item is not returnable. View return policy
Versatile, comfortable design provides dependable, all-day head protection
  • Grooved HDPE outer shell has rain trough and slots for accessories
  • Available in two replaceable nylon web suspension systems, each with three height adjustments
  • Four-point suspension with pin-lock adjustment is sturdy and easy to use
  • Four-point suspension with ratchet adjustment offers a firmer, more comfortable fit
  • When properly adjusted, ratchet headband keeps cap firmly on head, even when working in awkward, head-down positions
Compliance
Meet ANSI™ Z89.1-2003 for Type I helmets, Class E.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI Z8.1-2003; Type I; Class E.
Color Orange
Suspension Type Pin-Lock