Learn More
Honeywell™ The Peak A79 Hard Hats
Description
- Grooved HDPE outer shell has rain trough and slots for accessories
- Available in two replaceable nylon web suspension systems, each with three height adjustments
- Four-point suspension with pin-lock adjustment is sturdy and easy to use
- Four-point suspension with ratchet adjustment offers a firmer, more comfortable fit
- When properly adjusted, ratchet headband keeps cap firmly on head, even when working in awkward, head-down positions
Compliance
Meet ANSI™ Z89.1-2003 for Type I helmets, Class E.
Specifications
Specifications
| Certifications/Compliance | Meets ANSI Z8.1-2003; Type I; Class E. |
| Color | Yellow |
| Suspension Type | Pin-Lock |