Description
- 9-base polycarbonate hardcoated lenses provide 180° field of distortion-free vision; block 99.9% UV
- Soft nosepiece conforms to various nose shapes for nonslip fit
- Available with scratch-resistant hardcoat or Fog-Ban™ antifog coating
Compliance
Meet or exceed ANSI™ Z87.1-2010 standards.
Specifications
Specifications
| Certifications/Compliance | Meets or exceeds ANSI Z87.1-2003 standards. |
| Lens Tint | Silver Mirror, Indoor/Outdoor |
| Frame Color | Gray |
| Lens Material | Polycarbonate |