Description
- Sporty, eight-base wraparound dual lens provides style, comfort and protection
- Temples with padded inserts offer a comfortable fit
- Available with scratch-resistant hardcoat or Fog-Ban™ antifog coating
- Non-slip rubber nosepiece
- Black frames
Compliance
Meets ANSI™ Z87.1-2010 standards.
Specifications
Specifications
| Lens Coating | Hardcoat |
| Certifications/Compliance | Meets ANSI Z87.1-2003 standards. |
| Lens Tint | Silver Mirror |
| Frame Color | Black |
| Lens Material | Polycarbonate |