Learn More
Honeywell North Adaptec™ Safety Eyewear
Catalog No. 19185697 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Color:
Black
Catalog No. Frame Color
19-185-697 Black
1 options
This item is not returnable. View return policy
Catalog No. 19-185-697 Supplier Honeywell Supplier No. T5900LTKSHS
  • $51.40
    $28.65 / Each
    Save $22.75
    44% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $2,231.00
    $1,433.65 / Case of 50 EA
    Save $797.35
    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Engineered to meet the demands of today's most diverse workforce offering 3 sizes including narrow, regular and wide to fit a wide variety of facial profiles and features. Delivers excellent protection, superior comfort, customized adjustability, premium optics and style
  • Three-position nose bridge is simple and easy to adjust and keeps eyes centered in the lens where optical clarity is best to decrease eyes strain and fatigue and increase comfort and productivity
  • Soft, flexible nose pad conforms to the face delivering greater comfort, contact and stability
  • Ratcheting temple hinges adjust lens angle for greater protection and reduced fogging
  • Propriertary temple design adjusts temple tension for personal comfort and non-slip security
  • Temple inserts slide from front to back for firmer or looser fit
  • Superior Hydroshield anti-fog coating keeps glasses consistently fog-free for 90X longer than the majority of other anti-fog products and 2X greater scratch resistance than our next best anti-fog coating
  • Easy, simple lens replacement system extends life of eyewear and lowers cost of ownership
Specifications
Lens Coating Anti-fog
Lens Type Hydroshield Anti-Fog
Shape Wraparound
Lens Tint Smoke
Product Type Safety Glasses
Frame Color Black
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Half Frame