Learn More
Honeywell™ North Adaptec™ Safety Glasses
Catalog No. 17986717
Change view
Click to view available options
Description:
Narrow Size, Dark Blue Frame, Clear Lens
Narrow Size, Dark Blue Frame, Smoke Lens
Narrow Size, Replacement Lens, Clear Lens
Narrow Size, Translucent-Black Frame, Amber Lens
Narrow Size, Translucent-Black Frame, I/O Mirror Lens
Narrow Size, Translucent-Black Frame, Smoke Lens
Regualr Size, Replacement Lens, Clear Lens
Regular Size, Black Frame, Amber Lens
Regular Size, Dark Blue Frame, Clear Lens
Regular Size, Translucent-Black Frame, Amber Lens
Regular Size, Translucent-Black Frame, Clear Lens
Regular Size, Translucent-Black Frame, I/O Mirror Lens
Regular Size, Translucent-Black Frame, IR Shade 3.0 Lens
Wide Size, Black Frame, Amber Lens
Wide Size, Black Frame, Clear Lens
Wide Size, Black Frame, Smoke Lens
Wide Size, Dark Blue Frame, Clear Lens
Wide Size, Dark Blue Frame, Smoke Lens
Wide Size, Replacement Lens, Clear Lens
Wide Size, Translucent-Black Frame, Clear Lens
Wide Size, Translucent-Black Frame, IR Shade 5.0 Lens
Wide Size, Translucent-Black Frame, Smoke Lens
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
17-986-717 Regular Size, Translucent-Black Frame, IR Shade 3.0 Lens
19-185-728 Wide Size, Black Frame, Smoke Lens
19-185-729 Wide Size, Black Frame, Amber Lens
19-185-735 Regular Size, Black Frame, Amber Lens
19-039-972 Wide Size, Black Frame, Clear Lens
17-986-712 Regular Size, Translucent-Black Frame, Clear Lens
17-986-711 Regular Size, Dark Blue Frame, Clear Lens
17-986-716 Regular Size, Translucent-Black Frame, I/O Mirror Lens
17-986-715 Regular Size, Translucent-Black Frame, Amber Lens
17-986-721 Narrow Size, Dark Blue Frame, Smoke Lens
17-986-719 Narrow Size, Dark Blue Frame, Clear Lens
17-986-722 Narrow Size, Translucent-Black Frame, Smoke Lens
17-986-723 Narrow Size, Translucent-Black Frame, Amber Lens
17-986-724 Narrow Size, Translucent-Black Frame, I/O Mirror Lens
17-986-727 Wide Size, Dark Blue Frame, Clear Lens
17-986-730 Wide Size, Translucent-Black Frame, Smoke Lens
17-986-728 Wide Size, Translucent-Black Frame, Clear Lens
17-986-729 Wide Size, Dark Blue Frame, Smoke Lens
17-986-734 Wide Size, Translucent-Black Frame, IR Shade 5.0 Lens
19-185-738 Wide Size, Replacement Lens, Clear Lens
19-185-739 Narrow Size, Replacement Lens, Clear Lens
19-185-740 Regualr Size, Replacement Lens, Clear Lens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
This item is not returnable. View return policy
Catalog No. 17-986-717 Supplier Honeywell™ Supplier No. T5900LTK3.0
This item is not returnable. View return policy
Honeywell™ North Adaptec™ Safety Glasses deliver excellent protection, superior comfort, customized adjustability, premium optics and stylish look.
  • Adjustable nose bridge centers eyes to reduce fatigue
  • Self-adjusting nose pads provde greater comfort, contact and stability
  • Vented cushioned frame allows optimum air circulation to reduce fogging
  • Temple hinge ratchets to seven positions — 30° of adjustment— to angle lens angle for optimal eye protection
  • Temple design adjusts tension for personal comfort
  • 99.9% UV protection
  • Replaceable lenses
  • Adjustable temples for comfort
  • Available in many lens tints, including SCT
Compliance
Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Anti-static, Scratch-resistant
Certifications/Compliance Meets ANSI Z87.1-2010, CSA Z94.3
Description Regular Size, Translucent-Black Frame, IR Shade 3.0 Lens
Product Type Safety glasses
Frame Color Translucent Black
Frame Material Polycarbonate
Frame Type Regular