Learn More
Honeywell™ North Adaptec™ Uvex HydroShield™ Anti-Fog Coating Safety Glasses
Catalog No. 19185696 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Description:
Narrow Size, Black/Red Frame, Amber Lens
Narrow Size, Black/Red Frame, Clear Lens
Narrow Size, Black/Red Frame, Smoke Lens
Regular Size, Black/Red Frame, Clear Lens
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
19-185-696 Regular Size, Black/Red Frame, Clear Lens
19-185-699 Narrow Size, Black/Red Frame, Clear Lens
19-185-700 Narrow Size, Black/Red Frame, Smoke Lens
19-185-761 Narrow Size, Black/Red Frame, Amber Lens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-185-696 Supplier Honeywell™ Supplier No. T5900LTKHS
Add to Cart
Add to Cart
Honeywell™ North Adaptec™ Safety Glasses deliver excellent protection, superior comfort, customized adjustability, premium optics and stylish look.
  • Adjustable nose bridge centers eyes to reduce fatigue
  • Self-adjusting nose pads provde greater comfort, contact and stability
  • Vented cushioned frame allows optimum air circulation to reduce fogging
  • Temple hinge ratchets to seven positions — 30° of adjustment— to angle lens angle for optimal eye protection
  • Temple design adjusts tension for personal comfort
  • 99.9% UV protection
  • Replaceable lenses
  • Adjustable temples for comfort
  • HydroShield Anti-Fog coating
Compliance
Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
Specifications
Lens Coating Anti-fog, Anti-static, Scratch-resistant
Certifications/Compliance Meets ANSI Z87.1-2010, CSA Z94.3
Description Regular Size, Black/Red Frame, Clear Lens
Lens Tint Clear
Product Type Safety Glasses
Frame Color Black/Red
Frame Material Polycarbonate
Frame Type Standard