Learn More
Honeywell™ North™ Adhesive Strip Bandages
Description
- Designed with a strong adhesive to adhere securely and protect the wound
- Island dressing provides a secure seal around the wound to help prevent contamination
Specifications
Specifications
| Product Type | Strip Bandage |
| For Use With (Application) | For a wide range of wound care applications |
| Adhesive | Aggressive Adhesive |
| Color | Beige |
| Material | Fabric |
| Quantity | 100/Pk. |
| Style | Strip |
| Dimensions (L x W) | 0.75 x 3 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.