Learn More
Honeywell™ North™ BP1000 Series Backpack Adapter
Catalog No. 19040886 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Backpack Adapter for North Air-Purifying Respirators
Catalog No.
19-040-886 Backpack Adapter for North Air-Purifying Respirators
1 options
Catalog No. 19-040-886 Supplier Honeywell™ Supplier No. BP1002
Add to Cart
Add to Cart
Convert your half mask or full facepiece to a welding respirator with this welding attachment. Puts cartridges on the back, increasing service life and minimizing weight to the facepiece.
  • Cartridges are located away from the immediate contaminate area so they last longer
  • Connects to North 5500, 7700, 5400 and 7600 Series facepieces
  • Converts half masks and full facepieces into a BP1000 series respirator
  • Creates a low profile that fits easily under welding helmets and faceshields when used with half masks
  • Weight is balanced on the shoulders, limiting worker fatigue
  • NIOSH certified when used with approved masks and facepieces
Specifications
Model BP1002