Learn More
Honeywell™ North™ BP1000 Series Backpack Air-Purifying Respirator
Description
- Ideal for welding and other high-heat operations
- Meets NIOSH 42 CFR Part 84 standards
Convenient backpack design allows cartridges and filters to be located on the user's back and out of the work zone—ideal for environments with high heat or sparks, such as welding, coke and smelting operations. Simply attach the backpack to North 7700 half mask cartridge connectors, then fasten the appropriate cartridges or filters to the backpack. No modifications, tools, or extra parts are needed.
Complete assembly includes backpack adapter and half-mask. Backpack adapter can also be purchased separately (below) to convert existing North facepieces to BP1000 Series air-purifying respirators
Specifications
Specifications
| Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
| Size | Medium |
| For Use With (Application) | Welding and other high-heat operations |
| For Use With (Equipment) | North™ 5500, 7700, 5400 and 7600 Series facepieces |
| Includes | Backpack adapter |
| Model | BP1201M |