Learn More
Honeywell™ North Conspire™ Safety Eyewear
Catalog No. 19066200
Change view
Click to view available options
Lens Coating:
Anti-fog
Scratch-resistant
Lens Tint:
Amber
Clear
Silver Mirror
Silver, Indoor/Outdoor Mirror
TSR Gray
Frame Color:
Matte Amber
Matte Clear
Matte Gray
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint Frame Color
19-066-200 Scratch-resistant Clear Matte Clear
19-066-205 Anti-fog Clear Matte Clear
19-066-201 - TSR Gray Matte Gray
19-066-203 Scratch-resistant Silver Mirror Matte Gray
19-066-204 Scratch-resistant Silver, Indoor/Outdoor Mirror Matte Clear
19-066-202 - Amber Matte Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-066-200 Supplier Honeywell™ Supplier No. XV400
Add to Cart
Add to Cart
This item is not returnable. View return policy
North Signature Fit™ concept profile for adjustment free fit for wide range of faces
  • Provides 99.9% UV protection
  • Lightweight, 3threepiece full-wrap design
  • Shaped nosepiece and flexible temples
  • Dielectric with no metal parts
  • Eight-base single front with dual lenses
  • Available in lens tints with Hard coat and Fog-Ban™ Anti-Fog coatings
Applications:
Construction, manufacturing industries.
Compliance
Meets the ANSI™ Z87.1-2010 and and CSA Z94.3 standards.
Specifications
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Safety Eyewear
For Use With (Application) Construction, Manufacturing
Frame Color Matte Clear