Learn More
Honeywell™ North Conspire™ Safety Eyewear
Catalog No. 19066201
Change view
Click to view available options
Lens Tint:
Amber
Clear
Silver Mirror
Silver, Indoor/Outdoor Mirror
TSR Gray
Frame Color:
Matte Amber
Matte Clear
Matte Gray
Lens Coating:
Anti-fog
Scratch-resistant
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Frame Color Lens Coating
19-066-201 TSR Gray Matte Gray -
19-066-205 Clear Matte Clear Anti-fog
19-066-200 Clear Matte Clear Scratch-resistant
19-066-203 Silver Mirror Matte Gray Scratch-resistant
19-066-204 Silver, Indoor/Outdoor Mirror Matte Clear Scratch-resistant
19-066-202 Amber Matte Amber -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-066-201 Supplier Honeywell™ Supplier No. XV401
Add to Cart
Add to Cart
This item is not returnable. View return policy
North Signature Fit™ concept profile for adjustment free fit for wide range of faces
  • Provides 99.9% UV protection
  • Lightweight, 3threepiece full-wrap design
  • Shaped nosepiece and flexible temples
  • Dielectric with no metal parts
  • Eight-base single front with dual lenses
  • Available in lens tints with Hard coat and Fog-Ban™ Anti-Fog coatings
Applications:
Construction, manufacturing industries.
Compliance
Meets the ANSI™ Z87.1-2010 and and CSA Z94.3 standards.
Specifications
Lens Tint TSR Gray
Product Type Safety Eyewear
For Use With (Application) Construction, Manufacturing
Frame Color Matte Gray