Learn More
Honeywell™ North Conspire™ Safety Eyewear
Description
- Provides 99.9% UV protection
- Lightweight, 3threepiece full-wrap design
- Shaped nosepiece and flexible temples
- Dielectric with no metal parts
- Eight-base single front with dual lenses
- Available in lens tints with Hard coat and Fog-Ban™ Anti-Fog coatings
Applications:
Construction, manufacturing industries.
Compliance
Meets the ANSI™ Z87.1-2010 and and CSA Z94.3 standards.
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Lens Tint | Silver, Indoor/Outdoor Mirror |
| Product Type | Safety Eyewear |
| For Use With (Application) | Construction, Manufacturing |
| Frame Color | Matte Clear |