Learn More
Honeywell™ North Conspire™ Safety Eyewear
Catalog No. 19066205
Change view
Click to view available options
Lens Coating:
Anti-fog
Scratch-resistant
Lens Tint:
Amber
Clear
Silver Mirror
Silver, Indoor/Outdoor Mirror
TSR Gray
Frame Color:
Matte Amber
Matte Clear
Matte Gray
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint Frame Color
19-066-205 Anti-fog Clear Matte Clear
19-066-201 - TSR Gray Matte Gray
19-066-200 Scratch-resistant Clear Matte Clear
19-066-203 Scratch-resistant Silver Mirror Matte Gray
19-066-204 Scratch-resistant Silver, Indoor/Outdoor Mirror Matte Clear
19-066-202 - Amber Matte Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-066-205 Supplier Honeywell™ Supplier No. XV405
  • $7.60
    $5.65 / Each
    Save $1.95
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $75.87
    $56.65 / Pack of 10 EA
    Save $19.22
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
North Signature Fit™ concept profile for adjustment free fit for wide range of faces
  • Provides 99.9% UV protection
  • Lightweight, 3threepiece full-wrap design
  • Shaped nosepiece and flexible temples
  • Dielectric with no metal parts
  • Eight-base single front with dual lenses
  • Available in lens tints with Hard coat and Fog-Ban™ Anti-Fog coatings
Applications:
Construction, manufacturing industries.
Compliance
Meets the ANSI™ Z87.1-2010 and and CSA Z94.3 standards.
Specifications
Lens Coating Anti-fog
Lens Type Uvextra Anti-Fog
Lens Tint Clear
Product Type Safety Eyewear
For Use With (Application) Construction, Manufacturing
Frame Color Matte Clear