Learn More
Honeywell™ North™ P100 Particulate Filter Accessory, Shower Cap
Catalog No. 19816208 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Shower Cap
Catalog No.
19-816-208 Shower Cap
1 options
This item is not returnable. View return policy
Catalog No. 19-816-208 Supplier Honeywell™ Supplier No. N750029
  • $9.98
    $6.65 / Each of 1
    Save $3.33
    33% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Shower Cap for P100 Filter protects filter from water spray during abatement and decontamination, as well as sparks and slag during welding and grinding operations. For use with 5500, RU8500 and 7700 Series half masks and 5400, RU6500 and 7600 Series full facepieces.
NIOSH certified when used with Honeywell and North facepieces.
Specifications
Product Type Shower Cap
For Use With (Application) Protects filter from water spray during abatement or during decontamination
For Use With (Equipment) 7580p100 Filter