Learn More
Honeywell™ North™ Primair™ 100 Series PA101 Loose-Fitting Facepiece Assembly
Loose-fitting assemblies are designed for pharmaceutical and other light industries.
Supplier:  Honeywell™ PA101M
 View more versions of this product
Catalog No. 19-134-241
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.
View Alternate Product
Includes: Loose-fitting facepiece assemblies include adjustable headgear, facepiece, and a replacement sweatband. Hood assemblies include adjustable headgear, hood, sweatband, and peel-off cover lens.
Description
Description
  • Spun-bound polypropylene hood cover with Velcro™ strip and rust-resistant snaps attaches and removes easily from headgear
  • Triple adjustment headgear provides a secure and comfortable fit
  • Replaceable 3/16 in. (0.48 cm) foam sweatband with adhesive backing adheres and removes easily from headband
  • Lens made of 0.040 mil co-polyester
  • Polypropylene chin cover
  • Hoods with collar and full bib have an assigned protection factor (APF) of 1000
  • Manifold directs air over lens to reduce fogging
Note: At the current time, the color of product received may be yellow instead of the white material as seen in the photo above.  
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25
Polypropylene
Medium
Compact Air 200 Series PAPR and CF1000 Series
NIOSH
Facepiece Assembly
Pharmaceutical and light duty applications
PA101M
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.