Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Compact Air Powered Air-Purifying Respirators: Primair Facepiece

Catalog No. 19134238 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Medium Replacement Primair Facepiece, Pk of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-134-238 Medium Replacement Primair Facepiece, Pk of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-134-238 Supplier Honeywell™ Supplier No. PA102M
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Replacement hood for Primair™ 101M hood assembly is a loose fitting facepiece with an under the chin, nylon collar. Size medium, 10/BX. Hoods have an assigned protection factor (APF) of 1000.

The Primair™ 100 Series includes one loose fitting head cover and several hoods that can be used with the CA200 Series PAPR and the CF1000 Series CF-SAR. The manifold directs air over lens to reduce fogging. Triple adjustment headgear for a comfortable fit.

Specifications

Color White
Material Tychem
Product Type Facepiece Assembly
For Use With (Equipment) Compact air 200 series powered air purifying respirator and cf1000 series
Model PA102M
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.