Learn More
Honeywell North™ Disposable Safety Packs
Catalog No. 19085569
Change view
Click to view available options
Includes:
Surgical Mask (1), Gloves (1 Pair), Sanitizing Wipes (2)
Surgical Mask (1), Gloves (1 Pair), Sanitizing Wipes (2), Ear Plugs (1 Pair)
Surgical Masks (3), Gloves (3 Pair), Sanitizing Wipes (5)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-085-569 Surgical Mask (1), Gloves (1 Pair), Sanitizing Wipes (2), Ear Plugs (1 Pair)
19-085-568 Surgical Mask (1), Gloves (1 Pair), Sanitizing Wipes (2)
19-085-570 Surgical Masks (3), Gloves (3 Pair), Sanitizing Wipes (5)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-085-569 Supplier Honeywell Supplier No. SAFETYPACK/PD/01
Packaged in an easy-to-open resealable pouch to provide employees, visitors, and consumers with easy access to safety products
  • Honeywell Safety Packs
  • Packaged in an easy-to-open, resealable pouch
  • Provides employees, visitors, and consumers with easy access to safety products
  • Surgical Mask: 3-ply, pleated
  • Gloves: Latex-free, powder-free
  • Sanitizing Wipes: 17 x 19 cm
  • Earplugs: Cordless, 32 db (NRR)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Safety Kit
Includes Surgical Mask (1), Gloves (1 Pair), Sanitizing Wipes (2), Ear Plugs (1 Pair)
Sterility Non-sterile
Disposable Single-use