Learn More
Honeywell™ North™ ER1000 Escape Respirator
Catalog No. 191202808
Change view
Click to view available options
ER1000 Escape Hood
Catalog No.
19-120-2808 ER1000 Escape Hood
1 options
Catalog No. 19-120-2808 Supplier Honeywell™ Supplier No. ER1000
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Ziplock storage bag allows wearer to practice donning prior to use
Disposable hood-style respirator provides emergency eye, face, and respiratory protection from NBC warfare agents and toxic industrial materials. Utilizes proven half-mask design for optimum comfort and communication ability. Offers more than 30 minutes of protection against high concentrations of various challenge agents including Sarin (GB) and mustard vapor (HD). Universal size. Single-use only.
  • Translucent polyurethane hood with scratch-resistant, antifog faceshield for high visibility and face recognition
  • ASZM-TEDA carbon impregnated Defend Plus cartridge with a HEPA particulate filter
  • Chemical-resistant butyl neck dam
  • Five-year shelf life in resealable foil storage bag