Learn More
Honeywell™ North™ Everest and Peak Series Hard Hats
Catalog No. 19999117
Change view
Click to view available options
Color:
Green
Orange
Red
Sky Blue
White
Yellow
Suspension Type:
6-Point Nylon Suspension w/3-level Height Adjustment
6-Point; 2-level Height Ratchet Adjustment
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-999-117 Yellow 6-Point; 2-level Height Ratchet Adjustment
19-999-114 Orange 6-Point Nylon Suspension w/3-level Height Adjustment
19-999-119 Red 6-Point Nylon Suspension w/3-level Height Adjustment
19-055-165 Green 6-Point Nylon Suspension w/3-level Height Adjustment
19-999-120 Sky Blue 6-Point Nylon Suspension w/3-level Height Adjustment
19-999-121 Yellow 6-Point Nylon Suspension w/3-level Height Adjustment
19-042-005 White 6-Point Nylon Suspension w/3-level Height Adjustment
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-999-117 Supplier Honeywell™ Supplier No. A49R020000
Full brim HDPE shell with accessory slots. Protects from UV rays, rain and falling debris. 4-point or 6-point nylon suspension with 2 level height adjustment. Meets ANSI Z89.1-2009 and CSA Z94.1-2005 Type 1 Class E.
  • Ideal for mining, forestry, utilities, the oil industry and other demanding environments
  • Accessory slots make this the first wide brim hard hat to accept face and hearing protection accessories for the fullest available protection
  • Choice of high quality suspensions designed for comfort, with forehead comfort band and crown pad, 2-level height adjustment and self-centering
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI Type 1; Class E
Color Yellow
Material HDPE
Suspension Type 6-Point; 2-level Height Ratchet Adjustment