Learn More
Honeywell™ North™ Fuse™ Safety Glasses
Deliver exceptional coverage with both style and comfort
Specifications
Frame Color Gun Metal
Includes Break-Away Flexicord
Lens Material Polycarbonate
View More Specs
Products 3
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity & Availability  
19-130-0839
View Documents
Honeywell™
11150800
Scratch-resistant Clear N/A N/A N/A
19-130-0843
View Documents
Honeywell™
11150806
Scratch-resistant TSR Gray N/A N/A N/A
19-130-0841
View Documents
Honeywell™
11150804
Scratch-resistant Silver Mirror N/A N/A N/A
Description
Description
Deliver exceptional coverage with both style and comfort
  • Lightweight, durable, gunmetal frame
  • Dual 9–base wrap-around lens design for full unobstructed field of vision
  • Adjustable saddle nose pads, soft temple tips
  • Durable polycarbonate lenses with scratch-resistant or Fog-Ban™ antifog coating
  • Adjustable breakaway Flexicord™ included
Compliance
Meet ANSI Z87.1-2010 and CSA Z94.3 safety standards
Specifications
Specifications
Gun Metal
Break-Away Flexicord
Polycarbonate
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.