Learn More
Honeywell™ North™ Hardhat: Ratchet Suspension
Catalog No. 19041993 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Product Type:
4-Point Ratchet
Catalog No. Product Type
19-041-993 4-Point Ratchet
1 options
Catalog No. 19-041-993 Supplier Honeywell™ Supplier No. A79RS2
  • $39.89
    $27.65 / Each of 1
    Save $12.24
    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Replacement suspension for Peak A79R Hard Hat with 4-point, nylon web suspension with ratchet size adjustment. Suspension features 3-level height adjustment.
The Peak A79R Hard Hat is a versatile and dependable design, with grooved HDPE outer shell, rain trough and accessory slots. Meets ANSI Z89.1-2009 and CSA Z94.1-2005 Type 1 Class E.
Specifications
Product Type 4-Point Ratchet
Suspension Type Ratchet