Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ HM500 Series Half Mask

Catalog No. 19185594 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-185-594 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-185-594 Supplier Honeywell™ Supplier No. HM501TS
  • $27.83
    $22.65 / Each

    Save $5.18

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $333.89
    $271.65 / Case of 12 EA

    Save $62.24

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Threaded half-mask is lightweight, durable and soft with easy operation features. Filters and cartridges not included.

  • Lighter than other similar masks with cartridges available on the market
  • Softer material for better touch and seal with skin, for long-time use
  • Professional design for universal fit
  • Advanced over-molding manufacturing and design made for intuitive assembly and disassembly
  • Drop-down function replaces need to doff hardhat or earmuff for quick work break (Fisher cat #'s 19-185-597, 598 and 599)
  • Easy cleaning and maintenance

Specifications

Connection Type Threaded
Color Black
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small
With Exhalation Valve Yes
For Use With (Application) Oil and gas, pharmaceutical, general manufacturing, metal industry, construction
For Use With (Equipment) N-Series filters and cartridges
Latex Free Yes
Model HM501TS
Sterility Non-sterile
Strap Attachment Type Non drop-down
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.